missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(2,3-dimethylphenyl)benzamide, TRC

Product Code. 30350557
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350557 1 g 1g
30354727 500 mg 500mg
30355107 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350557 Supplier Toronto Research Chemicals Supplier No. TRCC5641201G
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196617-88-6
Molecular Formula C15H14ClNO
Molecular Weight (g/mol) 259.73
Synonym Mefenamic RC2, NSC 164235
IUPAC Name 2-chloro-N-(2,3-dimethylphenyl)benzamide
SMILES Cc1cccc(NC(=O)c2ccccc2Cl)c1C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C15H14ClNO/c1-10-6-5-9-14(11(10)2)17-15(18)12-7-3-4-8-13(12)16/h3-9H,1-2H3,(H,17,18)
Chemical Name or Material 2-Chloro-N-(2,3-dimethylphenyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.