missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-N-(2,6-dimethylphenyl)acetamide, 98%

Product Code. 11357200 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11357200 10 g Glass bottle 10g
10366490 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11357200 Supplier Thermo Scientific Acros Supplier No. 135440100
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1131-01-7 | C10H12ClNO | 197.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-01-7
Molecular Formula C10H12ClNO
Molecular Weight (g/mol) 197.66
InChI Key FPQQSNUTBWFFLB-UHFFFAOYSA-N
Synonym 2-chloro-n-2,6-dimethylphenyl acetamide, 2-chloro-2',6'-dimethylacetanilide, n-chloroacetyl-2,6-dimethylaniline, 2-chloro-2',6'-acetoxylidide, chloroaceto-2,6-xylidide, acetamide, 2-chloro-n-2,6-dimethylphenyl, chloroacetamido-2,6-xylidine, 1-chloroacetylamino-2,6-dimethylbenzene, n-2,6-dimethylphenyl-2-chloroacetamide, unii-5d2b69oapx
PubChem CID 70798
IUPAC Name 2-chloro-N-(2,6-dimethylphenyl)acetamide
SMILES CC1=C(C(=CC=C1)C)NC(=O)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 143.0°C to 148.0°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula (CH3)2C6H3NHC(=O)CH2Cl
Quantity 10 g
Beilstein 12,III,2464
Solubility Information Solubility in water: insoluble
Formula Weight 197.66
Percent Purity 98%
Physical Form Crystalline Powder or Needles
Chemical Name or Material 2-Chloro-N-(2, 6-dimethylphenyl)acetamide
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-N-(2,6-dimethylphenyl)acetamide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.