missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(hydroxymethyl)acetamide, TRC

Product Code. 30350533
Change view
Click to view available options
Quantity:
2.5 g
5 g
10 g
Unit Size:
2.5g
5g
Product Code. Quantity unitSize
30350533 2.5 g 2.5g
30355504 10 g -
30359080 5 g 5g
3 options
This item is not returnable. View return policy

Product Code. 30350533

Brand: Toronto Research Chemicals TRCC6701502.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2832-19-1
Molecular Formula C3H6ClNO2
Molecular Weight (g/mol) 123.54
Synonym Chloro-N-(hydroxymethyl)acetamide, N-(Hydroxymethyl)-2-chloracetamide, N-(Hydroxymethyl)-2-chloroacetamide, N-(Hydroxymethyl)chloroacetamide, N-Methylol-2-chloroacetamide, N-Methylol-alpha-chloracetamide, N-Methylol-alpha-chloroacetamide, N-Methylolchloracetamide, N-Methylolchloroacetamide, NSC 9963, alpha-Chloro-N-(hydroxymethyl)acetamide
IUPAC Name 2-chloro-N-(hydroxymethyl)acetamide
SMILES OCNC(=O)CCl

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C3H6ClNO2/c4-1-3(7)5-2-6/h6H,1-2H2,(H,5,7)
Formula Weight 123.0087
Chemical Name or Material 2-Chloro-N-(hydroxymethyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.