missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloro-N,N-diethylethyl-d4-amine HCl, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.05g
0.1g
Specifications
Specifications
| Chemical Name or Material | 2-Chloro-N,N-diethylethylamine 1,1,2,2-d4 hydrochloride |
| CAS | 1219805-94-3 |
| Molecular Formula | C6 2H4 H10 Cl N . Cl H |
| Quantity | 0.1 g |
| InChI Formula | InChI=1 S/C6H14ClN.ClH/c1-3-8(4-2)6-5-7;/h3-6H2,1-2H3;1 H/i5D2,6D2; |
| SMILES | Cl.[2 H]C([2 H])(Cl)C([2 H])([2 H])N(CC)CC |
| IUPAC Name | 2-chloro-1,1,2,2-tetradeuterio-N,N-diethylethanamine;hydrochloride |
| Molecular Weight (g/mol) | 176.12 |
| Formula Weight | 175.0833 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction