missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacetamide, 98%

Product Code. 10071660 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
1kg
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10071660 5 g Glass bottle 5g
10021580 500 g Plastic bottle 500g
10477730 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10071660 Supplier Thermo Scientific Acros Supplier No. 148410050
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 79-07-2 | C2H4ClNO | 93.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-07-2
Molecular Formula C2H4ClNO
Molecular Weight (g/mol) 93.51
MDL Number MFCD00008027
InChI Key VXIVSQZSERGHQP-UHFFFAOYSA-N
Synonym chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
PubChem CID 6580
IUPAC Name 2-chloroacetamide
SMILES NC(=O)CCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 116.0°C to 120.0°C
Color White
Density 0.8400g/mL
Boiling Point 224.5°C
Flash Point 170°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CONH2
Quantity 5 g
Beilstein 02, 199
Fieser 13,70
Merck Index 15, 2110
Specific Gravity 0.84
Solubility Information Solubility in water: 90g/L (20°C). Other solubilities: soluble in alcohol, insoluble in ether
Formula Weight 93.51
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroacetamide, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloroacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H361f-Suspected of damaging fertility.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.