missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacetamide, TRC

Product Code. 30137281
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30137281 10 g 10g
30133677 50 g 50g
30003471 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137281 Supplier Toronto Research Chemicals Supplier No. TRCC35350010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-07-2
Formula Weight 92.9981
InChI Formula InChI=1S/C2H4ClNO/c3-1-2(4)5/h1H2,(H2,4,5)
IUPAC Name 2-chloroacetamide
Molecular Formula C2 H4 Cl N O
Molecular Weight (g/mol) 93.51
SMILES NC(=O)CCl
Synonym Acetamide, α-chloro- (4CI), 2-Chloroacetamide, Chloracetamide, Chloroacetamide, KM 101, KM 101 (fungicide), Mergal AF, Microcide, NSC 54286, NSC 8408, α-Chloroacetamide
Chemical Name or Material 2-Chloroacetamide

Specifications

Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.