missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetophenone, 97%

Product Code. 10062410 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10062410 25 g Glass bottle 25g
10397880 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10062410 Supplier Thermo Scientific Acros Supplier No. 179770250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2142-68-9 | C8H7ClO | 154.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2142-68-9
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.6
MDL Number MFCD00000560
InChI Key ZDOYHCIRUPHUHN-UHFFFAOYSA-N
Synonym 2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene
PubChem CID 72864
IUPAC Name 1-(2-chlorophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Amber to Yellow
Density 1.1880g/mL
Boiling Point 228.0°C to 229.0°C
Flash Point 88°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4COCH3
Refractive Index 1.5420 to 1.5450
Quantity 25 g
Beilstein 07, I, 151
Specific Gravity 1.188
Formula Weight 154.6
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2'-Chloroacetophenone, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.