missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacrylic acid, 96%, stab. with ca 0.3% BHT

Product Code. 11310014
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11310014 2 g 2g
11320014 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11310014 Supplier Thermo Scientific Alfa Aesar Supplier No. L00350.04
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 598-79-8 | C3H3ClO2 | 106.51 g/mol

2-Chloroacrylic acid is used as a halochemicals, organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroacrylic acid is used as a halochemicals, organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-79-8
Molecular Formula C3H3ClO2
Molecular Weight (g/mol) 106.51
MDL Number MFCD00014336
InChI Key SZTBMYHIYNGYIA-UHFFFAOYSA-N
Synonym 2-chloroacrylic acid, chloroacrylic acid, 2-propenoic acid, 2-chloro, alpha-chloroacrylic acid, acrylic acid, 2-chloro, 2-chloro-2-propenoic acid, .alpha.-chloroacrylic acid, acmc-1aqt3
PubChem CID 11735
ChEBI CHEBI:73963
IUPAC Name 2-chloroprop-2-enoic acid
SMILES OC(=O)C(Cl)=C

Specifications

Melting Point 63°C to 67°C
Quantity 2 g
UN Number UN3261
Beilstein 1098503
Solubility Information Soluble in water.
Formula Weight 106.51
Percent Purity ≥98%
Chemical Name or Material 2-Chloroacrylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.