missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroaniline-4,6-d2, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 2-Chloroaniline-4,6-d2 |
| CAS | 347840-10-2 |
| Molecular Formula | C6 D2 H4 Cl N |
| Quantity | 0.5 g |
| Synonym | Benzen-2,4-d2-amine, 6-chloro- (9 CI), 2-Chloroaniline-4,6-d2, 2-Chloroaniline-4,6-D2, 2-Chloroaniline-d2, 6-Chlorobenzen-2,4-d2-amine, 2-Chlorobenzen-4,6-d2-amine |
| InChI Formula | InChI=1 S/C6H6ClN/c7-5-3-1-2-4-6(5)8/h1-4 H,8H2/i1D,4 D |
| SMILES | [2 H]c1cc([2 H])c(N)c(Cl)c1 |
| IUPAC Name | 2-chloro-4,6-dideuterioaniline |
| Molecular Weight (g/mol) | 129.584 |
| Formula Weight | 129.031 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction