missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline hydrochloride, 97%

Product Code. 11363097
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11363097 25 g 25g
11373097 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363097 Supplier Thermo Scientific Alfa Aesar Supplier No. L13483.14
 Restricted Item
Estimated Shipment: 08-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 137-04-2 | C6H7Cl2N | 164.03 g/mol

2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-04-2
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.03
MDL Number MFCD00053282
InChI Key DRGIDRZFKRLQTE-UHFFFAOYSA-N
Synonym 2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1
PubChem CID 67298
IUPAC Name 2-chloroaniline;hydrochloride
SMILES [H+].[Cl-].NC1=CC=CC=C1Cl

Specifications

Melting Point ∼220°C (sublimation)
Odor Characteristic
Quantity 25 g
UN Number UN2018
Beilstein 3624280
Solubility Information It is soluble in water.
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material 2-Chloroaniline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.