missing translation for 'onlineSavingsMsg'
Learn More

2-chlorobenzene-1-sulfonyl chloride, Thermo Scientific™

Product Code. 11306276
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11306276 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11306276 Supplier Thermo Scientific Maybridge Supplier No. RF02645EA
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2905-23-9
Density 1.5480g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H4Cl2O2S
Linear Formula ClC6H4SO2Cl
MDL Number MFCD00051520
Quantity 10 g
Synonym 2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
InChI Key KMVZDSQHLDGKGV-UHFFFAOYSA-N
SMILES ClC1=CC=CC=C1S(Cl)(=O)=O
IUPAC Name 2-chlorobenzenesulfonyl chloride
Molecular Weight (g/mol) 211.06
PubChem CID 76187
Formula Weight 211.07
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.