missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzeneboronic acid, 97%

Product Code. 11460937
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11460937 5 g 5g
11450937 1 g 1g
11470937 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460937 Supplier Thermo Scientific Alfa Aesar Supplier No. B23324.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3900-89-8 | C6H6BClO2 | 156.37 g/mol

2-Chlorobenzeneboronic acid is used in the preparation of imidazo[1,2-a]pyridine amides which has tuberculostatic activity. It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobenzeneboronic acid is used in the preparation of imidazo[1,2-a]pyridine amides which has tuberculostatic activity. It is used as pharmaceutical intermediates.

Solubility
Soluble in DMSO, Methanol.

Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3900-89-8
Molecular Formula C6H6BClO2
Molecular Weight (g/mol) 156.37
MDL Number MFCD00674012
InChI Key RRCMGJCFMJBHQC-UHFFFAOYSA-N
Synonym 2-chlorophenyl boronic acid, 2-chlorobenzeneboronic acid, 2-chlorophenylboronicacid, 2-chlorophenylbornic acid, o-chlorophenylboronic acid, 2-chlorophenyl boranediol, boronic acid, 2-chlorophenyl, 2-chloro-phenyl-boronic acid, boronic acid, b-2-chlorophenyl
PubChem CID 2734322
IUPAC Name (2-chlorophenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1Cl

Specifications

Melting Point ∼95°C to 100°C
Quantity 5 g
Beilstein 3030414
Solubility Information Soluble in DMSO,Methanol.
Formula Weight 156.38
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.