missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl mercaptan, 97%

Product Code. 11492498
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492498 5 g 5g
11402508 25 g 25g
11412508 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11492498 Supplier Thermo Scientific Alfa Aesar Supplier No. A15826.06
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 39718-00-8 | C7H7ClS | 158.643 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39718-00-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00004868
InChI Key WWFIIZLHSNBNTC-UHFFFAOYSA-N
Synonym 2-chlorobenzenemethanethiol, 2-chlorophenyl methanethiol, 2-chlorobenzyl mercaptan, 2-chlorobenzylmercaptan, 2-chloro benzyl mercaptan, benzenemethanethiol, 2-chloro, o-chlorobenzylmercaptan, pubchem6836, 2-chlorophenyl-methanethiol, d0p3xw
PubChem CID 580759
IUPAC Name (2-chlorophenyl)methanethiol
SMILES C1=CC=C(C(=C1)CS)Cl

Specifications

Density 1.22
Quantity 5 g
Refractive Index 1.591
UN Number UN2810
Beilstein 2042183
Boiling Point 120°C to 122°C (25 mmHg)
Formula Weight 158.65
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzyl mercaptan

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.