missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutane, 98+%

Product Code. 11427194
Change view
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427194 250 mL 250mL
11369639 50 mL 50mL
11417194 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427194 Supplier Thermo Scientific Alfa Aesar Supplier No. B21586.AK
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-86-4 | C4H9Cl | 92.566 g/mol

2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

Solubility
Immiscible with water.

Notes
Incompatible with metals, copper, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-86-4
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000871
InChI Key BSPCSKHALVHRSR-UHFFFAOYSA-N
Synonym sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl
PubChem CID 6563
IUPAC Name 2-chlorobutane
SMILES CCC(C)Cl

Specifications

Melting Point °C to 141 to°C to 140°C
Density 0.873
Boiling Point 67°C to 69°C
Flash Point -15°C (5°F)
Assay Percent Range 98+%
Refractive Index 1.397
Quantity 250 mL
UN Number UN1127
Beilstein 1718770
Merck Index 14,1561
Solubility Information Immiscible with water.
Formula Weight 92.57
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Chlorobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.