missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutane, 99+%

Product Code. 10696672 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10696672 250 mL Glass bottle 250mL
10204330 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10696672 Supplier Thermo Scientific Acros Supplier No. 108832500
  • ONLINE EXCLUSIVE PRICE
    37.50 €
    34.50 € / 250mL
    Save 3.00 €
    8% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 78-86-4 | C4H9Cl | 92.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-86-4
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00000871
InChI Key BSPCSKHALVHRSR-UHFFFAOYSA-N
Synonym sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl
PubChem CID 6563
IUPAC Name 2-chlorobutane
SMILES CCC(C)Cl

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -140.0°C
Color Colorless to Yellow
Density 0.8700g/mL
Boiling Point 68.0°C to 70.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C2H5CH(Cl)CH3
Refractive Index 1.3960 to 1.3980
Quantity 250 mL
Beilstein 01, 119
Merck Index 15, 1562
Specific Gravity 0.87
Solubility Information Solubility in water: 1g/L (25°C)
Formula Weight 92.57
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Chlorobutane, 99+%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.