missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutane, Extra Pure, SLR, Fisher Chemical

Product Code. 10293750 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10293750 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10293750 Supplier Fisher Chemical Supplier No. C/4759/05
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 78-86-4
Melting Point -140°C
Boiling Point 68°C
Packaging Amber glass bottle
Molecular Formula C4H9Cl
MDL Number 871
Quantity 100 mL
Formula Weight 92.57g/mol
Vapor Pressure 145mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorobutane
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.