missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutyric acid, 97%, Thermo Scientific Chemicals

Product Code. 11344516
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11344516

Brand: Thermo Scientific Alfa Aesar L20083.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4170-24-5
Molecular Formula C4H7ClO2
Molecular Weight (g/mol) 122.548
MDL Number MFCD00021697
InChI Key RVBUZBPJAGZHSQ-UHFFFAOYSA-N
Synonym 2-chlorobutyric acid, butanoic acid, 2-chloro, butyric acid, 2-chloro, ccris 8992, 2-chloro-butyric acid, 2-chlorbutyric acid, acmc-209huc, acmc-209lcl, 2-chloro-n-butyric acid, butanoic acid,2-chloro
PubChem CID 20137
IUPAC Name 2-chlorobutanoic acid
SMILES CCC(C(=O)O)Cl

Specifications

Density 1.19
Boiling Point 90°C to 92°C (12 mmHg)
Flash Point 112°C (233°F)
Refractive Index 1.439
Quantity 100 g
UN Number UN3265
Beilstein 1720944
Formula Weight 122.55
Percent Purity 97%
Chemical Name or Material 2-Chlorobutyric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.