missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chlorobutyryl chloride, tech. 85%

Product Code. 11397626
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397626 5 g 5g
11307636 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397626 Supplier Alfa Aesar™ Supplier No. L11207.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
2-Chlorobutyryl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7623-11-2
MDL Number MFCD00041409

Specifications

Density 1.236
Boiling Point 138°C to 140°C
Molecular Formula C12H13NO2S
Refractive Index 1.451
Quantity 5 g
UN Number UN3265
Beilstein 741947
Sensitivity Moisture sensitive
Synonym ethyl 3-methyl-4h-benzo b 1,4 thiazine-2-carboxylate, ethyl 3-methyl-4h-benzo e 1,4-thiazine-2-carboxylate, 3-methyl-4h-benzo 1,4 thiazine-2-carboxylic acid ethyl ester, maybridge1_006434, cbmicro_028856, 2-carbethoxy-3-methyl-4h-1,4-benzothiazine, 3-methyl-4h-1,4-benzothiazine-2-carboxylic acid ethyl ester, 4h-1,4-benzothiazine-2-carboxylicacid, 3-methyl-, ethyl ester, 4h-1,4-benzothiazine-2-carboxylicacid,3-methyl-,ethyl ester
Solubility Information Reacts with water.
InChI Key NCZIRXVLKKERGP-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(NC2=CC=CC=C2S1)C
IUPAC Name ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate
Molecular Weight (g/mol) 235.30
PubChem CID 360308
Formula Weight 141
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Chlorobutyryl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.