missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclohexanone, 95%, stabilized

Product Code. 10752505 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10752505

Brand: Thermo Scientific Acros 108960050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 822-87-7 | C6H9ClO | 132.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-87-7
Molecular Formula C6H9ClO
Molecular Weight (g/mol) 132.59
MDL Number MFCD00001626
InChI Key CCHNWURRBFGQCD-UHFFFAOYNA-N
Synonym 2-chlorocyclohexanone, cyclohexanone, 2-chloro, alpha-chlorocyclohexanone, 2-chlorocyclohexonone, .alpha.-chlorocyclohexanone, 2-chlorocyclohexanon, 2-chlorocylohexanone, 2-chloro cyclohexanone, 2-chloro-cyclohexanone, acmc-209ted
PubChem CID 13203
IUPAC Name 2-chlorocyclohexan-1-one
SMILES ClC1CCCCC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 23°C
Color Brown
Density 1.1600g/mL
Boiling Point 82°C to 83°C (10.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic bottle
Linear Formula ClC6H9(=O)
Refractive Index 1.4830 to 1.485
Quantity 5 g
Beilstein 07, 10
Specific Gravity 1.16
Stabilizer 0.2% CaCO3/MgO (1:1)
Formula Weight 132.59
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chlorocyclohexanone, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorocyclohexanone, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.