missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclohexanone, 97%, stab. with calcium carbonate/magnesium oxide

Product Code. 11458537
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11458537

Brand: Thermo Scientific Alfa Aesar A18613.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 822-87-7 | C6H9ClO | 132.59 g/mol

2-Chlorocyclohexanone, is an intermediate in the synthesis of variety of pharmaceutical compounds. It is also an intermediate in the synthesis of 1-Chloro-2-methylenecyclohexane .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorocyclohexanone, is an intermediate in the synthesis of variety of pharmaceutical compounds. It is also an intermediate in the synthesis of 1-Chloro-2-methylenecyclohexane .

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents and heat. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep refrigerated at -20°C.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-87-7
Molecular Formula C6H9ClO
Molecular Weight (g/mol) 132.59
MDL Number MFCD00001626
InChI Key CCHNWURRBFGQCD-UHFFFAOYNA-N
Synonym 2-chlorocyclohexanone, cyclohexanone, 2-chloro, alpha-chlorocyclohexanone, 2-chlorocyclohexonone, .alpha.-chlorocyclohexanone, 2-chlorocyclohexanon, 2-chlorocylohexanone, 2-chloro cyclohexanone, 2-chloro-cyclohexanone, acmc-209ted
PubChem CID 13203
IUPAC Name 2-chlorocyclohexan-1-one
SMILES ClC1CCCCC1=O

Specifications

Melting Point 22°C to 24°C
Density 1.161
Boiling Point 82°C to 83°C (10 mmHg)
Flash Point 82°C (180°F)
Refractive Index 1.484
Quantity 50 g
Beilstein 774100
Solubility Information Insoluble in water.
Formula Weight 132.59
Percent Purity 97%
Chemical Name or Material 2-Chlorocyclohexanone, stabilized with calcium carbonate/magnesium oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.