missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclopentanone, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number 10102810
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
694-28-0
C5H7ClO
118.56
MFCD00001410
AXDZFGRFZOQVBV-UHFFFAOYNA-N
2-chlorocyclopentanone, cyclopentanone, 2-chloro, 2-chloro-1-cyclopentanone, alpha-chlorocyclopentanone, 2-chlorocyclopentan-one, .alpha.-chlorocyclopentanone, chlorocyclopentanone, o-chlorocyclopentanone, wln: l5vtj bg, acmc-209u9b
12751
2-chlorocyclopentan-1-one
ClC1CCCC1=O
This item is not returnable. View return policy
694-28-0
C5H7ClO
118.56
MFCD00001410
AXDZFGRFZOQVBV-UHFFFAOYNA-N
2-chlorocyclopentanone, cyclopentanone, 2-chloro, 2-chloro-1-cyclopentanone, alpha-chlorocyclopentanone, 2-chlorocyclopentan-one, .alpha.-chlorocyclopentanone, chlorocyclopentanone, o-chlorocyclopentanone, wln: l5vtj bg, acmc-209u9b
12751
2-chlorocyclopentan-1-one
ClC1CCCC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Density 1.1800g/mL
Boiling Point 72.0°C to 74.0°C (12.0 mmHg)
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClC5H7(=O)
Refractive Index 1.4730 to 1.4770
Quantity 5 g
Beilstein 07, I, 5
Specific Gravity 1.18
Stabilizer about 1% dry potassium carbonate
Formula Weight 118.56
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chlorocyclopentanone, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorocyclopentanone, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'