missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanesulfonyl chloride, 95%

Product Code. 10711442 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10711442 25 g Glass bottle 25g
10290103 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10711442 Supplier Thermo Scientific Acros Supplier No. 404610250
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1622-32-8 | C2H4Cl2O2S | 163.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1622-32-8
Molecular Formula C2H4Cl2O2S
Molecular Weight (g/mol) 163.02
MDL Number MFCD00007461
InChI Key VHCSBTPOPKFYIU-UHFFFAOYSA-N
Synonym 2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
PubChem CID 15385
IUPAC Name 2-chloroethanesulfonyl chloride
SMILES C(CCl)S(=O)(=O)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.5500g/mL
Boiling Point 78°C (9.0 mmHg)
Flash Point 99°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (ex Cl) (Argentometry)
Packaging Glass bottle
Linear Formula ClCH2CH2SO2Cl
Refractive Index 1.4910 to 1.494
Quantity 25 g
Specific Gravity 1.55
Formula Weight 163.02
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chloroethanesulfonyl chloride, 95%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloroethanesulfonyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H302-Harmful if swallowed.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.