missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Acidity | 0.2 mg KOH/g max. |
| CAS Min % | 99.0 |
| CAS Max % | 100.0 |
| Melting Point | -63.0°C |
| Color | Colorless |
| Density | 1.2000 g/mL |
| Boiling Point | 129.0°C |
| Flash Point | 55°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% min. (GC) |
| Show More |
Safety and Handling
Product Identifier
- 2-Chloroethanol
- Danger
- Flammable liquid Category 3
- Acute toxicity Category 2
- Acute toxicity Category 1
- H226-Flammable liquid and vapour.
- H330-Fatal if inhaled.
- H310-Fatal in contact with skin.
- H300-Fatal if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P361-Take off immediately all contaminated clothing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?