missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloroethyl)benzene, 99%

Product Code. 11321747
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11321747 10 g 10g
11331747 100 g 100g
11341747 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11321747 Supplier Thermo Scientific Alfa Aesar Supplier No. L04891.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 622-24-2 | C8H9Cl | 140.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 622-24-2
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000977
InChI Key MNNZINNZIQVULG-UHFFFAOYSA-N
Synonym 2-chloroethyl benzene, phenethyl chloride, 2-phenylethyl chloride, benzene, 2-chloroethyl, 1-chloro-2-phenylethane, phenethylchloride, ethylchlorobenzene, 2-phenyl-1-chloroethane, 2-chloroethyl-benzene, beta-phenethyl chloride
PubChem CID 231496
IUPAC Name 2-chloroethylbenzene
SMILES ClCCC1=CC=CC=C1

Specifications

Melting Point -60°C
Density 1.07
Boiling Point 82°C to 84°C (16 mmHg)
Flash Point 66°C (150°F)
Refractive Index 1.53
Quantity 10 g
Beilstein 385969
Formula Weight 140.61
Percent Purity 99%
Chemical Name or Material 2-(Chloroethyl)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.