missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl isocyanate, 98%

Product Code. 10607202 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
1mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10607202 1 mL Glass bottle 1mL
10368210 5 mL Glass bottle 5mL
10617202 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10607202 Supplier Thermo Scientific Acros Supplier No. 221190010
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1943-83-5 | C3H4ClNO | 105.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1943-83-5
Molecular Formula C3H4ClNO
Molecular Weight (g/mol) 105.52
MDL Number MFCD00002043
InChI Key BCMYXYHEMGPZJN-UHFFFAOYSA-N
Synonym 2-chloroethyl isocyanate, 2-chloroethylisocyanate, chloroethylisocyanate, ethane, 1-chloro-2-isocyanato, 2-chlorethylisokyanat, 2-chloro ethyl isocyanate, isocyanic acid, 2-chloroethyl ester, 1-chloro-2-isocyanato-ethane, 2-chlorethylisokyanat czech, .beta.-chloroethylisocyanate
PubChem CID 16035
IUPAC Name 1-chloro-2-isocyanatoethane
SMILES C(CCl)N=C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2370g/mL
Boiling Point 135°C
Flash Point 56°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH2NCO
Refractive Index 1.4460 to 1.449
Quantity 1 mL
Beilstein 04, III, 245
Specific Gravity 1.237
Solubility Information Solubility in water: reacts
Formula Weight 105.52
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl isocyanate, 98%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloroethyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.