missing translation for 'onlineSavingsMsg'
Learn More

(2-Chloroethyl)trimethylammonium chloride, 98% (dry wt.)

Product Code. 11497517
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497517 25 g 25g
11487517 5 g 5g
11407527 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497517 Supplier Thermo Scientific Alfa Aesar Supplier No. A15630.14
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 999-81-5 | C5H13Cl2N | 158.066 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 999-81-5
Molecular Formula C5H13Cl2N
Molecular Weight (g/mol) 158.066
MDL Number MFCD00011869
InChI Key UHZZMRAGKVHANO-UHFFFAOYSA-M
Synonym chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
PubChem CID 13836
IUPAC Name 2-chloroethyl(trimethyl)azanium;chloride
SMILES C[N+](C)(C)CCCl.[Cl-]

Specifica

Melting Point ∼241°C (decomposition)
Odor Amine-like
Linear Formula ClCH2CH2N(CH3)3Cl
Quantity 25 g
UN Number UN2811
Beilstein 3563994
Merck Index 14,2104
Formula Weight 158.07
Percent Purity 98%
Assay (dry wt.), may cont. up to 5% water
Chemical Name or Material (2-Chloroethyl)trimethylammonium chloride
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.