missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethylamine hydrochloride, 98+%

Product Code. 11417127
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417127 250 g 250g
11407127 50 g 50g
11497117 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11417127 Supplier Thermo Scientific Alfa Aesar Supplier No. A14455.30
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 870-24-6 | C2H7Cl2N | 115.99 g/mol

2-chloroethylamine hydrochloride is used as an intermediate in active pharmaceutical ingredients and dyes. It is also used in organic synthesis. Further, it is employed as a derivatizing reagent for amino acids, dipeptides and nucleotides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-chloroethylamine hydrochloride is used as an intermediate in active pharmaceutical ingredients and dyes. It is also used in organic synthesis. Further, it is employed as a derivatizing reagent for amino acids, dipeptides and nucleotides.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-24-6
Molecular Formula C2H7Cl2N
Molecular Weight (g/mol) 115.99
MDL Number MFCD00012887,MFCD00137399
InChI Key ONRREFWJTRBDRA-UHFFFAOYSA-N
Synonym 2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
PubChem CID 9793737
IUPAC Name 2-chloroethanamine;hydrochloride
SMILES [Cl-].[NH3+]CCCl

Specifications

Melting Point 143°C to 148°C
pH 2 to 3
Odor Odorless
Quantity 250 g
UN Number UN3261
Beilstein 3590304
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 115.99
Percent Purity ≥98%
Chemical Name or Material 2-Chloroethylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloroethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Chloroethylamine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.