missing translation for 'onlineSavingsMsg'
Learn More

2-chloroethylamine hydrochloride, 98%

Product Code. 10568853 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10568853 25 g Glass bottle 25g
10249990 100 g Glass bottle 100g
13478568 1 kg Plastic bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10568853 Supplier Thermo Scientific Acros Supplier No. 422860250
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 870-24-6 | C2H7Cl2N | 115.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 870-24-6
Molecular Formula C2H7Cl2N
Molecular Weight (g/mol) 115.99
MDL Number MFCD00012887,MFCD00137399
InChI Key ONRREFWJTRBDRA-UHFFFAOYSA-N
Synonym 2-chloroethylamine hydrochloride, 2-chloroethanamine hydrochloride, 1-amino-2-chloroethane hydrochloride, ethanamine, 2-chloro-, hydrochloride, 2-aminoethyl chloride hydrochloride, ethanamine, 2-chloro-, hydrochloride 1:1, 2-chloroethan-1-amine hydrochloride, chlorethamine hydrochloride, 2-chloro-ethylamine hydrochloride, chlorethamine hcl
PubChem CID 9793737
SMILES [Cl-].[NH3+]CCCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 140.0°C to 150.0°C
Color White to Beige
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCH2CH2NH2·HCl
Quantity 25 g
Beilstein 04,133
Solubility Information Solubility in water: soluble in water
IUPAC Name 2-chloroethan-1-aminium chloride
Formula Weight 115.99
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloroethylamine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloroethylamine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.