missing translation for 'onlineSavingsMsg'
Learn More

2-chloroethyleisothiocyanate, 98%, ACROS Organics™

Product Code. 10389310 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10389310 1g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10389310 Supplier Thermo Scientific Acros Supplier No. 309430010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 6099-88-3
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C3H4ClNS
Linear Formula ClCHCHNCS
Refractive Index 1.5550 to 1.5570 (20°C, 589nm)
MDL Number MFCD00041125
Quantity 1 g
Synonym 2-chloroethyl isothiocyanate, 2-chloroethylisothiocyanate, ethane, 1-chloro-2-isothiocyanato, sgxdlhbabrkth@, chloroethylisothiocyanate, acmc-1b2t4, beta-chloroethylisothiocyanate, beta-chloroethyl isothiocyanate, 1-chloro-2-isothiocyanato-ethane, #
InChI Key ZUWFBQUHBOUPFK-UHFFFAOYSA-N
SMILES C(CCl)N=C=S
IUPAC Name 1-chloro-2-isothiocyanatoethane
Molecular Weight (g/mol) 121.582
PubChem CID 138661
Formula Weight 121.59
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 2-chloroethyleisothiocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.