missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroimidazole, 97%

Product Code. 15478347
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15478347 250 mg 250mg
15458347 1 g 1g
15468347 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15478347 Supplier Thermo Scientific Alfa Aesar Supplier No. H51863.MD
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Aggiungi al carrello
Request Bulk or Custom Format
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 16265-04-6 | C3H3ClN2 | 102.52 g/mol

2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 16265-04-6
Molecular Formula C3H3ClN2
Molecular Weight (g/mol) 102.52
MDL Number MFCD02179530
InChI Key OCVXSFKKWXMYPF-UHFFFAOYSA-N
Synonym 2-chloroimidazole, chloroimidazole, sftheadililuh@, pubchem7597, 2-chlor-1h-imidazol, zlchem 1064, acmc-209dos, 1h-imidazole,2-chloro, 2-chloranyl-1h-imidazole, 1h-imidazole, 2-chloro
PubChem CID 2773328
ChEBI CHEBI:52721
IUPAC Name 2-chloro-1H-imidazole
SMILES ClC1=NC=CN1

Specifica

Melting Point 165°C to 166°C
Quantity 250 mg
Formula Weight 102.52
Percent Purity 97%
Chemical Name or Material 2-Chloroimidazole

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.