missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroisonicotinic Acid, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | 2-Chloroisonicotinic Acid |
| CAS | 6313-54-8 |
| Molecular Formula | C6H4ClNO2 |
| Quantity | 1 g |
| Synonym | 2-Chloro-4-pyridinecarboxylic Acid, 2-Chloropyridine-4-carboxylic Acid, 6-Chloroisonicotinic Acid, NSC 40139 |
| InChI Formula | InChI=1S/C6H4ClNO2/c7-5-3-4(6(9)10)1-2-8-5/h1-3H,(H,9,10) |
| SMILES | OC(=O)c1ccnc(Cl)c1 |
| IUPAC Name | 2-chloropyridine-4-carboxylic acid |
| Molecular Weight (g/mol) | 157.55 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction