missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-2-methyloxirane, 97%, Thermo Scientific™

Product Code. 10069301
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10069301 1 g Amber Glass Bottle 1g
10111122 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10069301 Supplier Thermo Scientific Maybridge Supplier No. CD08593DA
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 598-09-4
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.549
MDL Number MFCD00052487
InChI Key VVHFXJOCUKBZFS-UHFFFAOYSA-N
Synonym 2-chloromethyl-2-methyloxirane, oxirane, 2-chloromethyl-2-methyl, 2-chloromethyl-2-methyl-oxirane, 2-chloromethyl-1,2-epoxypropane, methyl epichlorohydrin, 2-methylepichlorohydrin, acmc-209mfx, beta-methylepichlorohydrin, ksc493o4b
PubChem CID 95220
IUPAC Name 2-(chloromethyl)-2-methyloxirane
SMILES CC1(CO1)CCl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 106.55
Percent Purity 97%
Chemical Name or Material 2-(Chloromethyl)-2-methyloxirane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.