missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine Hydrochloride, TRC

Product Code. 30365264
Change view
Click to view available options
Quantity:
2.5 g
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365264 2.5 g -
30352628 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30365264 Supplier Toronto Research Chemicals Supplier No. TRCC3692402.5G
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86604-75-3
Molecular Formula C9 H12 Cl N O . Cl H
Molecular Weight (g/mol) 222.11
Synonym 2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride (1:1), Pyridine, 2-(chloromethyl)-4-methoxy-3,5-dimethyl-, hydrochloride (9CI), 2-(Chloromethyl)-3,5-dimethyl-4-methoxypyridine hydrochloride, 2-(Chloromethyl)-4-methoxy-3,5-dimethylpyridine hydrochloride, 3,5-Dimethyl-2-chloromethyl-4-methoxypyridine hydrochloride, Cc-993
IUPAC Name 2-(chloromethyl)-4-methoxy-3,5-dimethylpyridine;hydrochloride
SMILES Cl.COc1c(C)cnc(CCl)c1C

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H12ClNO.ClH/c1-6-5-11-8(4-10)7(2)9(6)12-3;/h5H,4H2,1-3H3;1H
Formula Weight 221.0374
Chemical Name or Material 2-Chloromethyl-3,5-dimethyl-4-methoxypyridine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.