missing translation for 'onlineSavingsMsg'
Learn More

2-Chloromethyl-5-(4-methoxyphenyl)-1,3,4-oxadiazole, 98%, Thermo Scientific™

Product Code. 10657952 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10657952 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10657952 Supplier Thermo Scientific Acros Supplier No. 354710010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 24023-71-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point 91°C to 95°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H9ClN2O2
MDL Number MFCD01230030
Quantity 1 g
Synonym 2-chloromethyl-5-4-methoxyphenyl-1,3,4-oxadiazole, 4-5-chloromethyl-1,3,4-oxadiazol-2-yl anisole, 1,3,4-oxadiazole, 2-chloromethyl-5-4-methoxyphenyl, 2-chloromethyl-5-4-methoxyphenyl-1?3?4-oxadiazole, 2-chloromethyl-5-4-methoxy-phenyl-1,3,4 oxadiazole, 1,3,4-oxadiazole,2-chloromethyl-5-4-methoxyphenyl, 1-5-chloromethyl 1,3,4-oxadiazol-2-yl-4-methoxybenzene, 2-chloromethyl-5-4-methoxyphenyl-1,2,4-oxadiazole
InChI Key GEHIXSKXGCIKJJ-UHFFFAOYSA-N
SMILES COC1=CC=C(C=C1)C1=NN=C(CCl)O1
IUPAC Name 2-(chloromethyl)-5-(4-methoxyphenyl)-1,3,4-oxadiazole
Molecular Weight (g/mol) 224.64
PubChem CID 2734194
Formula Weight 224.65
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chloromethyl-5-(4-methoxyphenyl)-1, 3, 4-oxadiazole, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloromethyl-5-(4-methoxyphenyl)-1,3,4-oxadiazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.