missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)-5-(4-methylphenyl)-1,3,4-oxadiazole, ≥95%, Thermo Scientific™

Product Code. 10314732
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10314732 1 g 1g
10611693 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10314732 Supplier Thermo Scientific Maybridge Supplier No. RF02900DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 287197-95-9
Molecular Formula C10H9ClN2O
Molecular Weight (g/mol) 208.645
MDL Number MFCD00662318
InChI Key JDAULQMOFMANSL-UHFFFAOYSA-N
Synonym 2-chloromethyl-5-4-methylphenyl-1,3,4-oxadiazole, 5-chloromethyl-2-4-methylphenyl-1,3,4-oxadiazole, 1,3,4-oxadiazole, 2-chloromethyl-5-4-methylphenyl, 1,3,4-oxadiazole,2-chloromethyl-5-4-methylphenyl, 2-chloromethyl-5-p-tolyl-1,3,4 oxadiazole, 2-chloromethyl-5-p-tolyl-1,3,4-oxadiazole, 1,3,4 oxadiazole, 2-chloromethyl-5-p-tolyl
PubChem CID 736973
IUPAC Name 2-(chloromethyl)-5-(4-methylphenyl)-1,3,4-oxadiazole
SMILES CC1=CC=C(C=C1)C2=NN=C(O2)CCl

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 208.65
Percent Purity 95%
Chemical Name or Material 2-(Chloromethyl)-5-(4-methylphenyl)-1, 3, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.