missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)pyrimidine hydrochloride, Thermo Scientific™

Product Code. 10472570
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10472570 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10472570 Supplier Thermo Scientific Maybridge Supplier No. MAY00188DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 936643-80-0
Packaging Amber Glass Bottle
Molecular Formula C5H6Cl2N2
MDL Number MFCD09834372
Quantity 1 g
Synonym 2-chloromethyl pyrimidine hydrochloride, 2-chloromethyl pyrimidine hcl, 2-chloromethyl-pyrimidine hydrochloride, 2-chloromethyl pyrimidine, chloride, 2-chloro mehyl pyrimidine hcl, pyrimidine,2-chloromethyl-,hydrochloride, 2-chloromethyl pyrimidine hydrochloride salt, 2-chloromethyl pyrimidine hydrogen chloride, 2-chloromethyl-pyrimidine, monohydrochloride
InChI Key XLGVMJXAZRCTRU-UHFFFAOYSA-N
SMILES Cl.ClCC1=NC=CC=N1
IUPAC Name 2-(chloromethyl)pyrimidine;hydrochloride
Molecular Weight (g/mol) 165.02
PubChem CID 20025606
Formula Weight 165.02
Percent Purity 95%
Chemical Name or Material 2-(Chloromethyl)pyrimidine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.