missing translation for 'onlineSavingsMsg'
Learn More

2-(Chloromethyl)quinoline hydrochloride, 97%

Product Code. 11498046
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11498046 1 g 1g
11408056 5 g 5g
11418056 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498046 Supplier Thermo Scientific Alfa Aesar Supplier No. A14102.03
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3747-74-8 | C10H9Cl2N | 214.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3747-74-8
Molecular Formula C10H9Cl2N
Molecular Weight (g/mol) 214.089
MDL Number MFCD00012734
InChI Key WDETYCRYUBGKCE-UHFFFAOYSA-N
Synonym 2-chloromethyl quinoline hydrochloride, 2-chloromethylquinoline hydrochloride, 2-chloromethyl quinoline hcl, 2-chloromethyl quinolinehydrochloride, 2-chloromethyl-quinoline hydrochloride, quinoline, 2-chloromethyl-, hydrochloride, acmc-209it8, ksc494s0b, 2-chloromethyl quinoline-hcl, 2-chloromethyl quinoline.hcl
PubChem CID 3083823
IUPAC Name 2-(chloromethyl)quinoline;hydrochloride
SMILES C1=CC=C2C(=C1)C=CC(=N2)CCl.Cl

Specifications

Melting Point 184°C to 190°C
Quantity 1 g
Beilstein 3565832
Formula Weight 214.1
Percent Purity 97%
Chemical Name or Material 2-(Chloromethyl)quinoline hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.