missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinoyl chloride, 97%

Product Code. 10614195 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10614195 25 g Glass bottle 25g
10061284 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10614195 Supplier Thermo Scientific Acros Supplier No. 395790250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 49609-84-9 | C6H3Cl2NO | 176.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49609-84-9
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 176.00
MDL Number MFCD00051677
InChI Key RXTRRIFWCJEMEL-UHFFFAOYSA-N
Synonym 2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride
PubChem CID 2774541
IUPAC Name 2-chloropyridine-3-carbonyl chloride
SMILES ClC(=O)C1=CC=CN=C1Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 38°C to 39°C
Color Beige-Yellow to White
Boiling Point 89°C to 91°C (1.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 176
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 2-Chloronicotinoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloronicotinoyl chloride
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.