missing translation for 'onlineSavingsMsg'
Learn More

2-Chloronicotinoyl chloride, 97%

Product Code. 11438893
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438893 100 g 100g
11418893 5 g 5g
11428893 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11438893 Supplier Thermo Scientific Alfa Aesar Supplier No. A11048.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 49609-84-9 | C6H3Cl2NO | 176.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 49609-84-9
Molecular Formula C6H3Cl2NO
Molecular Weight (g/mol) 176.00
MDL Number MFCD00051677
InChI Key RXTRRIFWCJEMEL-UHFFFAOYSA-N
Synonym 2-chloronicotinoyl chloride, 2-chloro-nicotinoyl chloride, 3-pyridinecarbonyl chloride, 2-chloro, 2-chloronicotinylchloride, pubchem2599, acmc-209kgw, chloronicotinoyl chloride, 2-chloronicotinoylchloride, 2-chloro nicotinoyl chloride, 2-chloronicotinic acid chloride
PubChem CID 2774541
IUPAC Name 2-chloropyridine-3-carbonyl chloride
SMILES ClC(=O)C1=CC=CN=C1Cl

Specifications

Melting Point 38°C to 42°C
Boiling Point 98°C to 100°C (2 mmHg)
Quantity 100 g
UN Number UN3261
Beilstein 119022
Sensitivity Moisture sensitive
Formula Weight 176
Percent Purity ≥98%
Chemical Name or Material 2-Chloronicotinoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.