missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2-Chlorophenyl)diphenyl-d5-methanol (phenyl-d5), CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | (2-Chlorophenyl)diphenylmethanol-D5 (phenyl-D5) |
| CAS | 1219802-30-8 |
| Molecular Formula | C19 D5 H10 Cl O |
| Quantity | 0.5 g |
| Synonym | (2-Chlorophenyl)diphenyl-d5-methanol (phenyl-d5), (2-Chlorophenyl)diphenylmethanol-D5 (phenyl-D5), 2-Chloro-α-phenyl-α-(phenyl-d5)benzenemethanol, o-Chlorophenyldiphenylmethanol-d5, (2-Chlorophenyl)(phenyl)(phenyl-d5)methanol |
| InChI Formula | InChI=1 S/C19H15ClO/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14,21 H/i1D,3 D,4 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C(O)(c2ccccc2)c3ccccc3Cl |
| IUPAC Name | (2-chlorophenyl)-(2,3,4,5,6-pentadeuteriophenyl)-phenylmethanol |
| Molecular Weight (g/mol) | 299.8056 |
| Formula Weight | 299.1125 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction