missing translation for 'onlineSavingsMsg'
Learn More

(2-Chlorophenyl)methanesulfonyl chloride, ≥97%, Thermo Scientific™

Product Code. 10441504
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10441504 250 mg Amber Glass Bottle 250mg
10536863 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10441504 Supplier Thermo Scientific Maybridge Supplier No. MO07829CB
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 77421-13-7
Molecular Formula C7H6Cl2O2S
Molecular Weight (g/mol) 225.08
MDL Number MFCD04116369
InChI Key CHPZYFXSICSCNY-UHFFFAOYSA-N
Synonym 2-chlorophenyl methanesulfonyl chloride, 2-chloro-phenyl-methanesulfonyl chloride, chloro 2-chlorophenyl methyl sulfone, pubchem5500, acmc-1bfc0, 2-chlorobenzylsulfonyl chloride, benzenemethanesulfonylchloride, 2-chloro, 2-chloro-phenyl methanesulfonyl chloride, 2-chlorophenyl methane-sulfonyl chloride
PubChem CID 2757801
IUPAC Name (2-chlorophenyl)methanesulfonyl chloride
SMILES ClC1=CC=CC=C1CS(Cl)(=O)=O

Specifications

CAS Min % 97.0
Quantity 250 mg
Packaging Amber Glass Bottle
Assay Percent Range 97%
Formula Weight 225.09
Percent Purity 97%
Chemical Name or Material (2-Chlorophenyl)methanesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.