missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloropropane-d7, CDN
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | 2-Chloropropane-D7 |
| CAS | 55956-02-0 |
| Molecular Formula | C3 D7 Cl |
| Quantity | 5 g |
| Synonym | Propane-1,1,1,2,3,3,3-d7, 2-chloro- (9 CI), 1,1,1,2,3,3,3-Heptadeutero-2-chloropropane, 2-Chloropropane-d7, 2-Chloro-1,1,1,2,3,3,3-heptadeuteriopropane, Isopropyl-d7 chloride, Perdeuterioisopropyl chloride, 2-Chloropropane-D7, 2-Chloropropane-1,1,1,2,3,3,3-d7 |
| InChI Formula | InChI=1 S/C3H7Cl/c1-3(2)4/h3H,1-2H3/i1D3,2D3,3 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Cl)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-chloro-1,1,1,2,3,3,3-heptadeuteriopropane |
| Molecular Weight (g/mol) | 85.5838 |
| Formula Weight | 85.0676 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction