missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Chloropropionic acid, 94%

Product Code. p-7020789
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11329639

Brand: Thermo Scientific Alfa Aesar A12860.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-78-7 | C3H5ClO2 | 108.52 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-78-7
Molecular Formula C3H5ClO2
Molecular Weight (g/mol) 108.52
MDL Number MFCD00004224
InChI Key GAWAYYRQGQZKCR-UHFFFAOYNA-N
Synonym 2-chloropropionic acid, propanoic acid, 2-chloro, propionic acid, 2-chloro, alpha-chloropropionic acid, alpha-monochloropropionic acid, propionic acid, alpha-chloro, .alpha.-chloropropionic acid, propionic acid, .alpha.-chloro, chloropropionic acid, propanoic acid, chloro
PubChem CID 11734
IUPAC Name 2-chloropropanoic acid
SMILES CC(Cl)C(O)=O

Specifications

Melting Point -13°C
Density 1.262
Boiling Point 78°C to 80°C (11 mmHg)
Flash Point 101°C (213°F)
Refractive Index 1.435
Quantity 100 g
UN Number UN2511
Beilstein 1720259
Formula Weight 108.53
Percent Purity 94%
Chemical Name or Material (+/-)-2-Chloropropionic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.