missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropropionyl chloride, 95%

Product Code. 10023450 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
This item is not returnable. View return policy

Product Code. 10023450

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7623-09-8 | C3H4Cl2O | 126.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7623-09-8
Molecular Formula C3H4Cl2O
Molecular Weight (g/mol) 126.97
MDL Number MFCD00000862
InChI Key JEQDSBVHLKBEIZ-UHFFFAOYNA-N
Synonym 2-chloropropionyl chloride, propanoyl chloride, 2-chloro, alpha-chloropropionyl chloride, 2-chloropropionic acid chloride, 2-chloropropionylchloride, .alpha.-chloropropionyl chloride, 2-chloropropanyl chloride, 1,2dichloro-1-propanone, 2-chloro-propionylchloride, 1,2-dichloro-1-propanone
PubChem CID 111019
IUPAC Name 2-chloropropanoyl chloride
SMILES CC(C(=O)Cl)Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point -71.0°C
Color Colorless to Yellow
Density 1.3000g/mL
Boiling Point 109.0°C to 111.0°C
Flash Point 34°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH(Cl)COCl
Refractive Index 1.4390 to 1.4410
Quantity 100 mL
Beilstein 02, 248
Specific Gravity 1.3
Solubility Information Solubility in water: reacts
Formula Weight 126.97
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chloropropionyl chloride, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloropropionyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.