missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrazine, 98%

Product Code. 11499342
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499342 100 g 100g
11479342 5 g 5g
11489342 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499342 Supplier Thermo Scientific Alfa Aesar Supplier No. A10108.22
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14508-49-7 | C4H3ClN2 | 114.53 g/mol

2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropyrazine is used in cardiotonic, in biological, drug, flavoring and perfumery industry.

Solubility
Soluble in Methanol

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14508-49-7
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD00006124
InChI Key GELVZYOEQVJIRR-UHFFFAOYSA-N
Synonym chloropyrazine, pyrazine, chloro, pyrazine, 2-chloro, 2-chloro-pyrazine, 2-chloro pyrazine, chioropyrazine, chloropyrazin, chloro-pyrazine, 6-chloropyrazine, 2-chloropyrazine
PubChem CID 73277
IUPAC Name 2-chloropyrazine
SMILES ClC1=CN=CC=N1

Specifications

Density 1.284
Boiling Point 153°C to 154°C
Flash Point 57°C (134°F)
Refractive Index 1.537
Quantity 100 g
UN Number UN1993
Beilstein 107019
Solubility Information Soluble in Methanol
Formula Weight 114.54
Percent Purity 98%
Chemical Name or Material 2-Chloropyrazine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.