missing translation for 'onlineSavingsMsg'
Learn More

2-Chloropyrimidine, 98%

Product Code. 11498346
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
Unit Size:
100g
10g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11498346 25 g 25g
11309639 10 g 10g
11408356 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11498346 Supplier Thermo Scientific Alfa Aesar Supplier No. A15394.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1722-12-9 | C4H3ClN2 | 114.53 g/mol

2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4?-(1,1?-(5-(2-methoxyphenoxy)-[2,2?-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloropyrimidine is a useful reagent in the preparation of antivirals and other biologically active compounds. It was used in the synthesis of 4′-(1,1′-(5-(2-methoxyphenoxy)-[2,2′-bipyrimidine]-4,6-diyl)bis(1H-pyrazol-3,1-diyl)) dianiline fluorescent dye, biosensor for protein assay, 2-amino-4-heteroarylpyrimidines, cis- and trans-octahydropyrrolo[2,3]pyridine derivatives.

Solubility
Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1722-12-9
Molecular Formula C4H3ClN2
Molecular Weight (g/mol) 114.53
MDL Number MFCD00006060
InChI Key UNCQVRBWJWWJBF-UHFFFAOYSA-N
Synonym pyrimidine, 2-chloro, 2-chloro-pyrimidine, chloropyrimidine, 2-chloro pyrimidine, 2-chloro-1,3-diazine, 2-chlorpyrimidin, 2-chloro-pyrimindine, 2chloropyrimidine, chloro-pyrimidine, 2-chloropyrimidin
PubChem CID 74404
IUPAC Name 2-chloropyrimidine
SMILES ClC1=NC=CC=N1

Specifications

Melting Point 64°C to 67°C
Boiling Point 75°C to 76°C (10 mmHg)
Flash Point 98°C (208°F)
Quantity 25 g
UN Number UN2811
Beilstein 107171
Sensitivity Hygroscopic
Solubility Information Slightly soluble in water. 33.3 mg soluble in 1 mL of alcohol.
Formula Weight 114.54
Percent Purity 98%
Chemical Name or Material 2-Chloropyrimidine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.