missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothiophene, 97%, Thermo Scientific™

Product Code. 10635032 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10635032 25 mL Glass bottle 25mL
10489260 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10635032 Supplier Thermo Scientific Acros Supplier No. 168260250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 96-43-5
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.58
MDL Number MFCD00005421
InChI Key GSFNQBFZFXUTBN-UHFFFAOYSA-N
Synonym 2-thienyl chloride, thiophene, 2-chloro, 2-chloro thiophene, 2-chlorthiophen, chlorothiophene, thienyl chloride, 5-chlorothiophene, thiophene, chloro, sfpdadhhryszzp@, pubchem7376
PubChem CID 7299
IUPAC Name 2-chlorothiophene
SMILES C1=CSC(=C1)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -71.9°C
Color Colorless to Brown
Density 1.2800g/mL
Boiling Point 127.0°C to 129.0°C
Flash Point 22°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5470 to 1.5490
Quantity 25 mL
Beilstein 17, 32
Specific Gravity 1.28
Solubility Information Solubility in water: insoluble in water. Other solubilities: miscible with ethanol, ether, slightly soluble in chloroform
Formula Weight 118.58
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chlorothiophene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chlorothiophene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.