missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorothioxanthone, 99%

Product Code. 11499286
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499286 5 g 5g
11409296 25 g 25g
11419296 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499286 Supplier Thermo Scientific Alfa Aesar Supplier No. A18131.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 86-39-5 | C13H7ClOS | 246.71 g/mol

2-Chlorothioxanthone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorothioxanthone is used as pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-39-5
Molecular Formula C13H7ClOS
Molecular Weight (g/mol) 246.71
MDL Number MFCD00005067
InChI Key ZCDADJXRUCOCJE-UHFFFAOYSA-N
Synonym 2-chlorothioxanthone, 2-chloro-9h-thioxanthen-9-one, 9h-thioxanthen-9-one, 2-chloro, unii-rgs87t004b, 2-chlorothiaxanthone, 2-chlorothioxanthene-9-one, sandoray 1050, 2-chloro-thioxanthone, pubchem10692, 2-chloro-9-thioxanthone
PubChem CID 618848
IUPAC Name 2-chloro-9H-thioxanthen-9-one
SMILES ClC1=CC=C2SC3=CC=CC=C3C(=O)C2=C1

Specifications

Melting Point 151°C to 154°C
Flash Point 91°C (196°F)
Quantity 5 g
Solubility Information Partly miscible in water.
Formula Weight 246.72
Percent Purity 99%
Chemical Name or Material 2-Chlorothioxanthone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.