missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotoluene, 98%

Product Code. 11415627
Change view
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415627 1000 g 1000g
11379629 100 g 100g
11425627 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11415627 Supplier Thermo Scientific Alfa Aesar Supplier No. B23596.0B
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-49-8 | C7H7Cl | 126.58 g/mol

2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-49-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000562
InChI Key IBSQPLPBRSHTTG-UHFFFAOYSA-N
Synonym 2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
PubChem CID 7238
IUPAC Name 1-chloro-2-methylbenzene
SMILES CC1=CC=CC=C1Cl

Specifications

Melting Point -36°C
Density 1.082
Boiling Point 157°C to 159°C
Quantity 1000 g
Refractive Index 1.525
UN Number UN2238
Beilstein 1904175
Merck Index 14,2171
Solubility Information Slightly soluble in water.
Formula Weight 126.59
Percent Purity 99%
Chemical Name or Material 2-Chlorotoluene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.