missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorotoluene, 98%

Product Code. 11425627
Click to view available options
Quantity:
100 g
250 g
1000 g
Unit Size:
1000g
100g
250g
This item is not returnable. View return policy

Product Code. 11425627

Brand: Thermo Scientific Alfa Aesar B23596.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-49-8 | C7H7Cl | 126.58 g/mol

2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorotoluene is used for permanganate oxidation to give o-chlorobenzoic acid. It is used in the preparation of 3-chloro-4-methylcatechol and 4-chloro-3-methylcatechol.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-49-8
Molecular Formula C7H7Cl
Molecular Weight (g/mol) 126.58
MDL Number MFCD00000562
InChI Key IBSQPLPBRSHTTG-UHFFFAOYSA-N
Synonym 2-chlorotoluene, o-chlorotoluene, benzene, 1-chloro-2-methyl, o-tolyl chloride, ortho-chlorotoluene, toluene, o-chloro, 2-methylchlorobenzene, halso 99, 2-chloro-1-methylbenzene, 1-methyl-2-chlorobenzene
PubChem CID 7238
IUPAC Name 1-chloro-2-methylbenzene
SMILES CC1=CC=CC=C1Cl

Specifications

Melting Point -36°C
Boiling Point 157°C to 159°C
Quantity 250 g
UN Number UN2238
Beilstein 1904175
Merck Index 14,2171
Formula Weight 126.59
Percent Purity 99%
Chemical Name or Material 2-Chlorotoluene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.